missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 1.00 N, Ricca Chemical

Supplier:  Ricca Chemical Company 83001

 View more versions of this product

Catalog No. 8300-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
93.0,4.78
97.0,4.97
4 L
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Laboratory
7732-18-5,7664-93-9
1.00N
Natural Poly Bottle
H2O4S
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836

Chemical Identifiers

7732-18-5
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Accessories
Promotions

Promotions