missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 1.00 N, Ricca Chemical
Supplier: Ricca Chemical Company 83004
Specifications
| Sulfuric Acid | |
| 93.0,4.78 | |
| 97.0,4.97 | |
| 120 mL | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| Laboratory |
| 7732-18-5,7664-93-9 | |
| 1.00N | |
| Natural Poly Bottle | |
| H2O4S | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 |
Chemical Identifiers
| 7732-18-5 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |