missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.0200 N (N/50), Ricca Chemical

Supplier:  Ricca Chemical Company RI820016

 View more versions of this product

Catalog No. 820016


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
98.0,0.1
Natural Poly Bottle
H2O4S
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
0.0200N
7732-18-5,7664-93-9
100.0,0.1
500 mL
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Laboratory

Chemical Identifiers

7732-18-5
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions