missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.0125 Normal, Reagents
Spécifications
Spécifications
| pH | 2 |
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Cubitainer |
| Specific Gravity | 1.0004 |
| Quantity | 20 L |
| Physical Form | Liquid |
| Grade | Reagent |
| Chemical Name or Material | Sulfuric Acid, 0.0125 Normal |