missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.0018 Normal, Reagents

Numéro de catalogue. 01333907
Modifier l'affichage
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
01333907 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01333907 Fournisseur Labchem Inc Code fournisseur CS0302004A
Il en reste null

Sulfuric Acid, 0.0018 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Spécifications

pH 2.8
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0006
Quantity 4 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.0018 Normal