missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC DSS8AY

 View more versions of this product

Catalog No. DSS8AY


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S50μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Sulfur Oil Standard in Base Oil
7704-34-9 , 8042-47-5
0.0,99.99
Solid
50 μg/g
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
NINIDFKCEFEMDL-UHFFFAOYSA-N
Base Oil 20
Petrochemical Testing
100 mL
CHEBI:26833
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.0,99.99
S
MFCD00085316
Certified Reference Material
[S]
sulfur
32.06
5362487

Chemical Identifiers

7704-34-9
32.06
NINIDFKCEFEMDL-UHFFFAOYSA-N
CHEBI:26833
[S]
S
MFCD00085316
5362487
sulfur
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only