missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC DSS875Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| S | 75μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Sulfur Oil Standard in Base Oil | |
| 7704-34-9 , 8042-47-5 | |
| 0.0,99.99 | |
| Solid | |
| 75 μg/g | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| NINIDFKCEFEMDL-UHFFFAOYSA-N | |
| Base Oil 20 | |
| Petrochemical Testing | |
| 100 mL | |
| CHEBI:26833 |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.0,99.99 | |
| S | |
| MFCD00085316 | |
| Certified Reference Material | |
| [S] | |
| sulfur | |
| 32.06 | |
| 5362487 |
Chemical Identifiers
| 7704-34-9 | |
| 32.06 | |
| NINIDFKCEFEMDL-UHFFFAOYSA-N | |
| CHEBI:26833 | |
| [S] |
| S | |
| MFCD00085316 | |
| 5362487 | |
| sulfur |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only