Learn More
Sulfur in Kerosene standard solution, Specpure™, 1000μg/g (0.10%)
Supplier: Thermo Scientific Chemicals 040541AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sulfur in Kerosene standard solution | |
| Liquid | |
| UN1223 | |
| Specpure™ |
| 1000μg/g (0.10%) | |
| MFCD00804268 | |
| 100 mL |
Chemical Identifiers
| MFCD00804268 |
Safety and Handling
GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
P210-P280-P301+P310-P331-P370+P378q-P501c
H227-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only