Learn More
Sulfur in Isooctane standard solution, Specpure™, 5μg/g (0.0005%)
Supplier: Thermo Scientific Chemicals 045319AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| Sulfur in Isooctane standard solution | |
| Petrol-like | |
| UN1262 | |
| Specpure™ |
| 5μg/g (0.0005%) | |
| MFCD00804269 | |
| 100 mL |
Identifiants chimiques
| MFCD00804269 |
Sécurité et manipulation
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
missing translation for 'einecsNumber' : 208-759-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only