missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Diesel Fuel standard solution, Specpure™, 5μg/g (0.0005%)

Catalog No. AA45308AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45308AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45308AE Supplier Thermo Scientific Chemicals Supplier No. 045308AE
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD01095233

Specifications

Density 0.6 to 0.9 g/mL
Flash Point 69°C (156°F)
Physical Form Liquid
Color Brown
Boiling Point 171°C to 315°C
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Diesel Fuel standard solution, 5μg/g (0.0005%)
Hazard Category H227-H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
EINECSNumber 270-676-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only