missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfosalicylic Acid, 5% (w/v), Ricca Chemical
Specifications
Specifications
| Percent Purity | 5% |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 5%(w/v) |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Aqueous solution |
| Quantity | 500 mL |
| Chemical Name or Material | Sulfosalicylic Acid |