missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 3% (w/v), Ricca Chemical

Catalog No. 811532
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
811532 1 L Amber Glass Bottle
811516 500 mL Amber Glass Bottle
81151 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 811532 Supplier Ricca Chemical Company Supplier No. 811532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97-05-2
Molecular Formula C7H6O6S
Molecular Weight (g/mol) 218.179
InChI Key YCPXWRQRBFJBPZ-UHFFFAOYSA-N
Synonym 5-sulfosalicylic acid, sulfosalicylic acid, 5-sulphosalicylic acid, salicylsulfonic acid, benzoic acid, 2-hydroxy-5-sulfo, salicylic acid, sulfo, benzoic acid, 2-hydroxysulfo, sulphosalicylic acid, salicylic acid, 5-sulfo, 2-hydroxybenzoic-5-sulfonic acid
PubChem CID 7322
ChEBI CHEBI:68555
IUPAC Name 2-hydroxy-5-sulfobenzoic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O

Specifications

Percent Purity 3%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 3% (w/v)
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous solution
Quantity 1 L
Chemical Name or Material Sulfosalicylic Acid Solution