missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 3% w/v, Reagents

Catalog No. 01333937
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333937 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333937 Supplier Labchem Inc Supplier No. CS0143001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfosalicylic Acid, 3% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 5965-83-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2-Hydroxy-5-sulfobenzoic Acid
IUPAC Name water
SMILES O

Specifications

Color Colorless to Light Pink
pH <1
Boiling Point 100°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.0117
Percent Purity 3%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Sulfosalicylic Acid, 3% w/v