missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 10% w/v, Reagents

Catalog No. p-200021621
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333952 1 L
01333951 4 L
01333950 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01333952 Supplier Labchem Inc Supplier No. CS0129001C
Only null left

Sulfosalicylic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 5965-83-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2-Hydroxy-5-sulfobenzoic Acid
IUPAC Name water
SMILES O

Specifications

Color Colorless to Light Pink
pH <1
Boiling Point 101°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.0446
Percent Purity 10% w/v
Grade Reagent
Physical Form Liquid
Chemical Name or Material Sulfosalicylic Acid, 10% w/v