missing translation for 'onlineSavingsMsg'
Learn More

Sulfolane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012857
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012857 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012857 Supplier Merck Emd Millipore Supplier No. 889655MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfolane is an aprotic polar solvent, used in the separation of aromatic hydrocarbons from aliphatic hydrocarbons.

Specifications

CAS 126-33-0
Vapor Pressure 0.01 mmHg (20°C)
Vapor Density 4.2 (vs. air)
Density 1.261 g/mL (at 25°C)
Flash Point 177°C
Physical Form Neat
Linear Formula C4H8O2 S
MDL Number MFCD00005484
Melting Point 20°C to 26°C
Boiling Point 104°C (0.2 mmHg); 285°C
Synonym Tetrahydrothiophene 1,1-dioxide; Tetramethylene sulfone
Type Analytical Standard
Quantity 5 mL
Formula Weight 120.17
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less