missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulfobromophthalein sodium salt hydrate

Catalog No. AA4387814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4387814 25 g
AA4387803 1 g
AA4387806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4387814 Supplier Thermo Scientific Chemicals Supplier No. 04387814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71-67-0 | C20H8Br4Na2O10S2 | 837.99 g/mol

Sulfobromophthalein sodium salt hydrate is a reagent for the quantitative determination of proteins, a dye for liver diagnostics in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-67-0
Molecular Formula C20H8Br4Na2O10S2
Molecular Weight (g/mol) 837.99
MDL Number MFCD00150017
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
Synonym bromosulfophthalein, sulfobromophthalein, 3h bsp, 3h-bsp, d00ady
PubChem CID 102371197
ChEBI CHEBI:63827
IUPAC Name disodium;2-sulfo-4-[4,5,6,7-tetrabromo-1-(4-oxido-3-sulfophenyl)-3-oxo-2-benzofuran-1-yl]phenolate
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C(Br)C(Br)=C(Br)C(Br)=C12)C1=CC=C(O)C(=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Sensitivity Moisture sensitive
Merck Index 14,8954
Solubility Information Soluble in water.
Formula Weight 837.98 (Anhydrous)
Physical Form Powder
Chemical Name or Material Sulfobromophthalein sodium salt hydrate
EINECSNumber 200-761-0
RTECSNumber SM8395000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only