missing translation for 'onlineSavingsMsg'
Learn More

Sulfisoxazole 98.0+%, TCI America™

Catalog No. U009825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U009825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U009825G Supplier TCI America Supplier No. U009825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127-69-5
Molecular Formula C11H13N3O3S
Molecular Weight (g/mol) 267.303
MDL Number MFCD00003150
InChI Key NHUHCSRWZMLRLA-UHFFFAOYSA-N
Synonym sulfisoxazole, sulfafurazole, sulphafurazole, sulfisoxazol, sulfafurazol, sulfaisoxazole, sulfofurazole, sulfisoxasole, sulphaisoxazole, sulfadimethylisoxazole
PubChem CID 5344
ChEBI CHEBI:102484
IUPAC Name 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
SMILES CC1=C(ON=C1C)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 195°C
Color White
Quantity 25 g
Formula Weight 267.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sulfisoxazole
RTECSNumber WO9100000
TSCA Yes
Recommended Storage Refrigerator