missing translation for 'onlineSavingsMsg'
Learn More

Sulfathiazole 98.0+%, TCI America™

Catalog No. S0272500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0272500G 500 g
S027225G 25 g
S0272100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0272500G Supplier TCI America Supplier No. S0272500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72-14-0
Molecular Formula C9H9N3O2S2
Molecular Weight (g/mol) 255.31
MDL Number MFCD00005319
InChI Key JNMRHUJNCSQMMB-UHFFFAOYSA-N
Synonym sulfathiazole, sulphathiazole, sulfathiazol, sulfanilamidothiazole, 2-sulfanilamidothiazole, norsulfazole, thiazamide, 2-sulfonamidothiazole, norsulfasol, neostrepsan
PubChem CID 5340
ChEBI CHEBI:9337
IUPAC Name 4-amino-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide
SMILES NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 500 g
Formula Weight 255.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulfathiazole
RTECSNumber WP2360000
TSCA Yes