missing translation for 'onlineSavingsMsg'
Learn More

Sulfathiazole, ≥98.0%, MilliporeSigma™ Supelco™

Catalog No. 111015347
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015347 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015347 Supplier Merck Emd Millipore Supplier No. S9876100G
Only null left

Sulfathiazole is a broad-spectrum antibacterial drug, belonging to the class of sulfonamides, used in veterinary and human medication for the treatment of infections. It is also involved in promoting growth of livestock and fish.

Environmental and Pharmaceutical (small molecule)

Specifications

Packaging Poly Bottle
CAS 72-14-0
Physical Form Neat
Linear Formula C9H9N3O2S2
MDL Number MFCD00005319
Melting Point 200°C to 202°C
Synonym 2-Sulfanilamidothiazole; 4-Amino-N-(2-thiazolyl)benzenesulfonamide; N1-(2-Thiazolyl)sulfanilamide
Type Analytical Standard
Quantity 100 g
Formula Weight 255.32
Percent Purity ≥98.0%
Grade Analytical Standard
Show More Show Less