missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate Standard Stock Solution, Alcoholic, 1000 ppm SO42- in 30% (v/v) Alcohol, Ricca Chemical
$271.79 - $326.81
Chemical Identifiers
| CAS | 7778-80-5 |
|---|---|
| Molecular Formula | K2O4S |
| Molecular Weight (g/mol) | 174.25 |
| MDL Number | MFCD00011388 |
| InChI Key | OTYBMLCTZGSZBG-UHFFFAOYSA-L |
| PubChem CID | 24507 |
| ChEBI | CHEBI:32036 |
| IUPAC Name | dipotassium sulfate |
| SMILES | [K+].[K+].[O-]S([O-])(=O)=O |
Chemical Identifiers
| 7778-80-5 | |
| 174.25 | |
| OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
| CHEBI:32036 | |
| [K+].[K+].[O-]S([O-])(=O)=O |
| K2O4S | |
| MFCD00011388 | |
| 24507 | |
| dipotassium sulfate |
Specifications
| Natural Poly Bottle | |
| Liquid | |
| MFCD00011388 | |
| Approximately 0°C | |
| [K+].[K+].[O-]S([O-])(=O)=O | |
| 174.25 | |
| 24507 | |
| Sulfate Standard Stock Solution |
| 7778-80-5,64-17-5,67-56-1,7732-18-5 | |
| K2O4S | |
| Colorless | |
| OTYBMLCTZGSZBG-UHFFFAOYSA-L | |
| dipotassium sulfate | |
| 120 mL | |
| CHEBI:32036 |