missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate Standard Stock Solution, Alcoholic, 1000 ppm SO42- in 30% (v/v) Alcohol, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | Approximately 0°C |
| Quantity | 120 mL |
| Chemical Name or Material | Sulfate Standard Stock Solution |