missing translation for 'onlineSavingsMsg'
Learn More

Sulfate Standard Stock Solution, 1000 ppm SO42-, Ricca Chemical

Catalog No. 81094
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
81094 120 mL Natural Poly Bottle
810916 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81094 Supplier Ricca Chemical Company Supplier No. 81094

Chemical Identifiers

CAS 7778-80-5 , 7732-18-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm SO
CAS Min % 98.0, 0.18
CAS Max % 100.0, 0.18
Physical Form Liquid
Color Colorless
pH Approximately 7
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Sulfate Standard, Stock Solution