missing translation for 'onlineSavingsMsg'
Learn More

Sulfate Standard, Certified, 100ppm ±1ppm (1mL = 0.1mg SO4), LabChem™

$82.93 - $119.35

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC255001
View Documents
LabChem
LC255001
500 mL Poly Bottle
Each for $82.93
Only null left
 
LC255002
View Documents
LabChem
LC255002
1 L Poly Bottle
Each for $119.35
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
Na2O4S
MFCD00003504
24436
disodium sulfate

Specifications

Poly Bottle
99.985,0.015
1.35cSt
Na2O4S
Na2SO4
Colorless
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
500 mL
CHEBI:32149
Certified
7757-82-6 , 7732-18-5
Passes Test
Liquid
100 ppm ±1 ppm (1mL = 0.1mg SO4)
MFCD00003504
Soluble in water
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
24436
142.04
Sulfate Standard, 100ppm (1mL = 0.1mg SO4)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents