missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate Standard, Certified, 100ppm ±1ppm (1mL = 0.1mg SO4), LabChem™
Supplier: LabChem LC255001
Specifications
| Sulfate Standard | |
| Poly Bottle | |
| 99.985,0.015 | |
| 1.35cSt | |
| Na2O4S | |
| Na2SO4 | |
| Colorless | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| 500 mL | |
| CHEBI:32149 | |
| Certified |
| 100ppm (1mL = 0.1mg SO4) | |
| 7757-82-6 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 100 ppm ±1 ppm (1mL = 0.1mg SO4) | |
| MFCD00003504 | |
| Soluble in water | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| 24436 | |
| 142.04 |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| CHEBI:32149 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| 24436 | |
| disodium sulfate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature