missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate Standard, Certified, 100ppm ±1ppm (1mL = 0.1mg SO4), LabChem™
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 99.985, 0.015 |
| Identification | Passes Test |
| Viscosity | 1.35cSt |
| Physical Form | Liquid |
| Concentration | 100 ppm ±1 ppm (1mL = 0.1mg SO4) |
| Linear Formula | Na2SO4 |
| Color | Colorless |
| Solubility | Soluble in water |
| Quantity | 1 L |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |