missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, 98.0-101.5%, MilliporeSigma™ Supelco™

Chemical structure: sulfonamide

$145.04 - $735.09

Specifications

CAS 599-79-1
Physical Form Powder
Linear Formula C18H14N4O5 S
MDL Number MFCD00057363
Melting Point 260°C to 265°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012222
View Documents
Merck Emd Millipore
S088310G
10 g
Each for $145.04
Only null left
 
111012223
View Documents
Merck Emd Millipore
S088350G
50 g
Each for $448.01
Only null left
 
111012224
View Documents
Merck Emd Millipore
S0883100G
100 g
Each for $735.09
Only null left
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

599-79-1
C18H14N4O5 S
260°C to 265°C
Analytical Standard
98.0 to 101.5%
Powder
MFCD00057363
2-Hydroxy-5-{{4-[(2-pyridinylamino)sulfonyl]phenyl}azo}benzoic acid; 5-[4-(2-Pyridylsulfamoyl)phenylazo]salicylic acid; Salicylazosulfapyridine
398.39
Analytical Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'