missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, 98.0-101.5%, MilliporeSigma™ Supelco™

Catalog No. 111012222
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012222 10 g
111012223 50 g
111012224 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111012222 Supplier Merck Emd Millipore Supplier No. S088310G

Chemical structure: sulfonamide

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 599-79-1
Physical Form Powder
Linear Formula C18H14N4O5 S
MDL Number MFCD00057363
Melting Point 260°C to 265°C
Synonym 2-Hydroxy-5-{{4-[(2-pyridinylamino)sulfonyl]phenyl}azo}benzoic acid; 5-[4-(2-Pyridylsulfamoyl)phenylazo]salicylic acid; Salicylazosulfapyridine
Type Analytical Standard
Quantity 10 g
Formula Weight 398.39
Percent Purity 98.0 to 101.5%
Grade Analytical Standard
Show More Show Less