missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide Reagent, 1% (w/v) in 10% (v/v) Hydrochloric Acid, Ricca Chemical

Chemical Identifiers

CAS 63-74-1 , 7647-01-0 , 7732-18-5
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
Synonym sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
PubChem CID 5333
ChEBI CHEBI:45373
IUPAC Name 4-aminobenzenesulfonamide
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
810316
View Documents
N/A 500 mL Natural Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

63-74-1 , 7647-01-0 , 7732-18-5
172.202
sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
CHEBI:45373
C1=CC(=CC=C1N)S(=O)(=O)N
C6H8N2O2S
FDDDEECHVMSUSB-UHFFFAOYSA-N
5333
4-aminobenzenesulfonamide

Specifications

63-74-1 , 7647-01-0 , 7732-18-5
sulfanilamide, sulphanilamide, sulfamine, sulphonamide, p-aminobenzenesulfonamide, sulfonylamide, sulfonamide, p-aminobenzenesulfamide, bacteramid, streptasol
C1=CC(=CC=C1N)S(=O)(=O)N
172.202
CHEBI:45373
1% (w/v)
C6H8N2O2S
FDDDEECHVMSUSB-UHFFFAOYSA-N
4-aminobenzenesulfonamide
5333
10% (v/v) Hydrochloric Acid
Sulfanilamide Reagent
Videos
SDS
Documents

Documents