missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, 99.0- 101.0%, Spectrum™ Chemical

$242.47 - $26,159.97

Chemical Identifiers

CAS 63-74-1
Molecular Weight (g/mol) 172.21
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18614161
View Documents Promotion Details
N/A 100 g Amber Glass Bottle
Each for $242.47
Only null left
 
18614165
View Documents Promotion Details
N/A 500 g Amber Glass Bottle
Each for $640.06
Only null left
 
18614163
View Documents Promotion Details
N/A 2.5 kg Amber Glass Bottle
Each for $2,522.76
Only null left
 
18614162
View Documents Promotion Details
N/A 12 kg Amber Glass Bottle
Each for $9,137.38
Only null left
 
18614164
View Documents Promotion Details
N/A 50 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

63-74-1
172.21

Specifications

Solid
1
White
99 to 101%
Sulfanilamide
63-74-1
0.005
172.21
Ungraded
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.