missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfanilamide, 99.0- 101.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor SU13450KGBL
| Quantity | 50 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 63-74-1 | |
| 172.21 |
Specifications
| Sulfanilamide | |
| 63-74-1 | |
| 0.005 | |
| White | |
| 172.21 | |
| Ungraded |
| Solid | |
| 1 | |
| Amber Glass Bottle | |
| 50 kg | |
| 99 to 101% |