missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, 99.0- 101.0%, Spectrum™ Chemical

Catalog No. 18614161
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614161 100 g Amber Glass Bottle
18614165 500 g Amber Glass Bottle
18614163 2.5 kg Amber Glass Bottle
18614162 12 kg Amber Glass Bottle
18614164 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614161 Supplier Spectrum Chemical Mfg Cor Supplier No. SU134100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 63-74-1
Molecular Weight (g/mol) 172.21

Specifications

Physical Form Solid
CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Color White
Quantity 100 g
Percent Purity 99 to 101%
Grade Ungraded
Chemical Name or Material Sulfanilamide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.