missing translation for 'onlineSavingsMsg'
Learn More

Sulfamonomethoxine 93.0+%, TCI America™

Catalog No. S05925G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05925G Supplier TCI America Supplier No. S05925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1220-83-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.302
MDL Number MFCD00063466
InChI Key WMPXPUYPYQKQCX-UHFFFAOYSA-N
Synonym sulfamonomethoxine, sulfamonomethoxin, daimeton, sulfamonometoxina, sulfamonomethoxinum, 4-amino-n-6-methoxypyrimidin-4-yl benzenesulfonamide, 4-amino-n-6-methoxy-4-pyrimidinyl benzenesulfonamide, benzenesulfonamide, 4-amino-n-6-methoxy-4-pyrimidinyl, n1-6-methoxy-4-pyrimidinyl sulfanilamide, sulfamonomethoxinum inn-latin
PubChem CID 5332
ChEBI CHEBI:32164
IUPAC Name 4-amino-N-(6-methoxypyrimidin-4-yl)benzenesulfonamide
SMILES COC1=NC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 280.30
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sulfamonomethoxine
RTECSNumber WP0550000
TSCA No
Recommended Storage Refrigerator