missing translation for 'onlineSavingsMsg'
Learn More

Sulfalène 98,0+%, TCI America™

Catalog No. S09175G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
S09175G 5 g
S091725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09175G Supplier TCI America Supplier No. S09175G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 152-47-6
Formule moléculaire C11H12N4O3S
Poids moléculaire (g/mol) 280.302
Numéro MDL MFCD00437754
Clé InChI KXRZBTAEDBELFD-UHFFFAOYSA-N
Synonyme 4-Amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide, N-(3-Methoxypyrazin-2-yl)sulfanilamide
PubChem CID 9047
ChEBI CHEBI:32162
Nom de l’IUPAC 4-amino-N-(3-méthoxypyrazine-2-yl)benzènesulfonamide
SOURIRES COC1=NC=CN=C1NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Nom ou substance chimique Sulfalene
Point de fusion 178 °C
Couleur Blanc
Quantité 5 g
Poids de formule 280.30
Pourcentage de pureté ≥98,0% (HPLC,T)
Forme physique Poudre cristalline
EINECSNumber (9)-0767
RTECSNumber WP0175000
TSCA Non