missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Sulbutiamine | |
| 137°C | |
| C32H46N8O6S2 | |
| 25 g | |
| CKHJPWQVLKHBIH-KKTFQPMKSA-N | |
| [(Z)-4-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-3-[[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-(2-methylpropanoyloxy)pent-2-en-3-yl]disulfanyl]pent-3-enyl] 2-methylpropanoate | |
| 24739911 | |
| ≥98.0% (HPLC,T) |
| 3286-46-2 | |
| White | |
| MFCD01726427 | |
| Bisibuthiamine, O-Isobutyroylthiamine Disulfide | |
| CC1=NC=C(C(=N1)N)CN(C=O)C(=C(CCOC(=O)C(C)C)SSC(=C(C)N(CC2=CN=C(N=C2N)C)C=O)CCOC(=O)C(C)C)C | |
| 702.89 | |
| 702.89 | |
| Crystalline Powder |
Chemical Identifiers
| 3286-46-2 | |
| 702.89 | |
| CKHJPWQVLKHBIH-KKTFQPMKSA-N | |
| 24739911 | |
| CC1=NC=C(C(=N1)N)CN(C=O)C(=C(CCOC(=O)C(C)C)SSC(=C(C)N(CC2=CN=C(N=C2N)C)C=O)CCOC(=O)C(C)C)C |
| C32H46N8O6S2 | |
| MFCD01726427 | |
| Bisibuthiamine, O-Isobutyroylthiamine Disulfide | |
| [(Z)-4-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-3-[[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-(2-methylpropanoyloxy)pent-2-en-3-yl]disulfanyl]pent-3-enyl] 2-methylpropanoate |
Safety and Handling
EINECSNumber : (5)-3823
RTECSNumber : LQ2721000
TSCA : No
Recommended Storage : Refrigerator