missing translation for 'onlineSavingsMsg'
Learn More

Sulbutiamine 98.0+%, TCI America™

Catalog No. p-7136005
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09645G 5 g
S096425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09645G Supplier TCI America Supplier No. S09645G
Only null left

Chemical Identifiers

CAS 3286-46-2
Molecular Formula C32H46N8O6S2
Molecular Weight (g/mol) 702.89
MDL Number MFCD01726427
InChI Key CKHJPWQVLKHBIH-KKTFQPMKSA-N
Synonym Bisibuthiamine, O-Isobutyroylthiamine Disulfide
PubChem CID 24739911
IUPAC Name [(Z)-4-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-3-[[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-(2-methylpropanoyloxy)pent-2-en-3-yl]disulfanyl]pent-3-enyl] 2-methylpropanoate
SMILES CC1=NC=C(C(=N1)N)CN(C=O)C(=C(CCOC(=O)C(C)C)SSC(=C(C)N(CC2=CN=C(N=C2N)C)C=O)CCOC(=O)C(C)C)C

Specifications

Melting Point 137°C
Color White
Quantity 5 g
Formula Weight 702.89
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sulbutiamine
EINECSNumber (5)-3823
RTECSNumber LQ2721000
TSCA No
Recommended Storage Refrigerator