accessibility menu, dialog, popup

UserName

CAS RN 3286-46-2

IUPAC Name: (3E)-4-{N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamido}-3-{[(2E)-2-{N-[(4-amino-2-methylpyrimidin-5-yl)methyl]formamido}-5-[(2-methylpropanoyl)oxy]pent-2-en-3-yl]disulfanyl}pent-3-en-1-yl 2-methylpropanoate
Synonyms: sulbutiamine
Molecular Weight (g/mol): 702.89
Molecular Formula: C32H46N8O6S2
InChi Key: CKHJPWQVLKHBIH-GPAWKIAZSA-N
SMILES: CC(C)C(=O)OCC\C(SS\C(CCOC(=O)C(C)C)=C(/C)N(CC1=CN=C(C)N=C1N)C=O)=C(\C)N(CC1=CN=C(C)N=C1N)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results