missing translation for 'onlineSavingsMsg'
Learn More

Soudan Red B, ≥97% (HPLC), MilliporeSigma™ Supelco™

Numéro de catalogue. 111013912
Modifier l'affichage
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111013912 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111013912 Fournisseur Merck Emd Millipore Code fournisseur 5160225MG

Spécifications

CAS 3176-79-2
Linear Formula C24H20N4 O
MDL Number MFCD00021456
Melting Point 173°C to 175°C
Type Analytical Standard
Quantity 25 mg
Formula Weight 380.44
Percent Purity ≥97% (HPLC)
Grade Norme analytique