missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan III

Numéro de catalogue. AAA1831836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1831836 500 g
AAA1831814 25 g
AAA1831822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1831836 Fournisseur Thermo Scientific Chemicals Code fournisseur A1831836
Il en reste null

CAS: 85-86-9 | C22H16N4O | 352.397 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
MDL Number MFCD00003905
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym C.I. 26100; Solvent Red 23
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Spécifications

Melting Point ∼200°C (decomposition)
Beilstein 931185
Merck Index 14,8884
Formula Weight 352.4
Quantity 500 g
Chemical Name or Material Sudan III
Hazard Category Warning
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-638-4
RTECSNumber QK4250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only