missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose Solution, NMR Reference Standard, 2 mM in H2O:D2O (9:1) (99.9 atom % D), DSS 0.5 mM, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | NMR Tube Size: 3 mm 8 in. |
| Concentration | 2 mM in H2O:D2O (9:1) (99.9 atom% D) |
| Linear Formula | C12H22O11 |
| Quantity | 0.235 mL |
| Formula Weight | 342.30 |
| Grade | NMR Reference Standard |