missing translation for 'onlineSavingsMsg'
Learn More

Sucralose, NF, 98-102%, Spectrum™ Chemical

Catalog No. 18607073
Change view
Click to view available options
Quantité:
100 g
Emballage:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité Emballage
18607073 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607073 Supplier Spectrum Chemical Mfg Cor Supplier No. S1416100GM
Jusqu’à 300 $ de produits chimiques supplémentaires
View all promotions

missing translation for 'validMainframeMsg'

S1416, 56038-13-2, C12H19Cl3O8

Spectrum™ Chemical Sucralose, NF is used as an artificial sweetener in phamaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 56038-13-2
CAS % maximal 1
Résidus de calcination 0.007
Emballage Amber Glass Bottle
Formule moléculaire C12H19Cl3O8
Numéro MDL MFCD03648615
Quantité 100 g
Rotation spécifique +84.0° to +87.5°
Clé InChI BAQAVOSOZGMPRM-QBMZZYIRSA-N
SOURIRES OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl
Nom de l’IUPAC (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
Poids moléculaire (g/mol) 397.63
Pourcentage de pureté 98 to 102%
Niveau NF
Show More Show Less