missing translation for 'onlineSavingsMsg'
Learn More

Sucralose, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014619
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014619 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014619 Supplier Merck Emd Millipore Supplier No. 90984100MG

Sucralose is a derivative of sucrose, which is generally used as a non-nutritive sweetener in foods and beverages.

Specifications

CAS 56038-13-2
Physical Form Neat
Linear Formula C12H19 Cl3O8
MDL Number MFCD03648615
Synonym 1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl-4-chloro-4-deoxy-alpha-D-galactopyranoside; E955; Trichlorosucrose
Type Analytical Standard
Quantity 100 mg
Formula Weight 397.63
Percent Purity ≥98% (HPLC)
Grade Analytical Standard