missing translation for 'onlineSavingsMsg'
Learn More

Succinyl Chloride 95.0+%, TCI America™

Catalog No. S011025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S011025G 25 g
S0110250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S011025G Supplier TCI America Supplier No. S011025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 543-20-4
Molecular Formula C4H4Cl2O2
Molecular Weight (g/mol) 154.974
MDL Number MFCD00000749
InChI Key IRXBNHGNHKNOJI-UHFFFAOYSA-N
Synonym succinyl chloride, succinyl dichloride, succinic acid dichloride, succinoyl chloride, succinoyl dichloride, succinic chloride, 1,2-bis chlorocarbonyl ethane, butanedioyl chloride, unii-gdn09v9867, 1,2-ethanediylbis chloroformate
PubChem CID 10970
IUPAC Name butanedioyl dichloride
SMILES C(CC(=O)Cl)C(=O)Cl

Specifications

Color Yellow
Boiling Point 192°C
Quantity 25 g
UN Number 3265
Formula Weight 154.97
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Succinyl Chloride
RTECSNumber WN4900000
TSCA Yes