missing translation for 'onlineSavingsMsg'
Learn More

Succinimide 98.0+%, TCI America™

Catalog No. S010825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S010825G 25 g
S0108500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S010825G Supplier TCI America Supplier No. S010825G

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O

Specifications

Melting Point 125°C
Color White
Boiling Point 130°C
Quantity 25 g
Formula Weight 99.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Succinimide
EINECSNumber (5)-0120
RTECSNumber WN2200000
TSCA Yes