missing translation for 'onlineSavingsMsg'
Learn More

Succinic acid disodium salt, hexahydrate, >98%, MP Biomedicals™

Catalog No. ICN10292980
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10292980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10292980 Supplier MP Biomedicals Inc Supplier No. 0210292980

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-90-3
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
InChI Key ZDQYSKICYIVCPN-UHFFFAOYSA-L
Synonym disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277
PubChem CID 9020
ChEBI CHEBI:63675
IUPAC Name disodium;butanedioate
SMILES C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 100 g
Formula Weight 270.1
Percent Purity >98%
Physical Form Powder
Chemical Name or Material Succinic acid disodium salt, hexahydrate