Learn More
Student Thermometers
Catalog No. S8819
Change view
Click to view available options
Includes:
2 x 6 in. Plastic backing
Catalog No. Includes
S8819 2 x 6 in. Plastic backing
1 options
Catalog No. S8819 Supplier Customer Channels Group (Ccg) Supplier No. LER 0302
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Easy-to-read.
  • Measures Fahrenheit and Celsius temperature accurately
  • With 2 x 6 in. plastic backing
Theremometer cannot be used in boiling water. The maximum temperature use is 50°Celsius.
Order Info
Grades 3-8. Set of 10.
Specifications
Includes 2 x 6 in. Plastic backing
For Use With (Application) Measures Fahrenheit and Celsius temperature accurately