missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Student Organic and Inorganic Chemistry Molecular Model Set

Catalog No. S96540
Change view
Click to view available options
Height (English):
1.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
S96540 1.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96540 Supplier Eisco™ Supplier No. SET00604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Student Organic and Inorganic Chemistry Molecular Model Set lets students visualize three-dimensional molecule structures and understand molecular formulas.

Hard plastic palls of different colors represent atoms; each has holes at correct angles to be connected with flexible plastic bonds. Medium links represent single covalent bonds while long links can be used for double and triple covalent bonds.

  • Six black carbon (4 holes)
  • 14 White hydrogen (1 hole)
  • Two blue nitrogen (4 holes)
  • One blue nitrogen (5 holes)
  • Six red oxygen (2 holes)
  • Three yellow sulfur (one each: 2 holes, 4 holes, and 6 holes)
  • One purple phosphorus (5 holes)
  • Six green halogen (1 hole)
  • Three gray metal (1 hole)
  • Three gray metal (2 holes)
  • Two gray metal (3 holes)
  • One gray metal (4 holes)
  • One brown™ sp3 (4 holes)
  • One brown™ dsp3 (5 holes)
  • One brown™ d2sp3 (6 holes)
  • 28 mm Gray links (20)
  • 43 mm Gray links (10)
  • 28 mm Purple links (5)
  • Instruction manual

Specifications

Dimensions (L x W x H) 9.8 x 7.4 x 1.5 in. (25 x 19 x 4 cm)
Height (English) 1.5 in.
Height (Metric) 4 cm
Includes Instructional guide
Length (English) 9.8 in.
Length (Metric) 25 cm
Material ABS Plastic
Weight (English) 1.3 lb.
Weight (Metric) 0.59 kg
Width (English) 7.4 in.
Width (Metric) 19 cm
Product Type Inorganic/Organic Chemistry Molecular Model Set
Show More Show Less