missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Student Distillation Kit

Catalog No. S40725
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S40725 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S40725 Supplier Eisco™ Supplier No. DIST1SET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Liebig condenser (30 mm), Kjeldahl ball (50 mm), Boiling flask (500 ml), Erlenmeyer flask (500 ml)support stand, wire gauze, support stand with rod, clamp with bosshead

This Student Distillation Kit contains everything needed for a simple distillation demonstration. Ideal for student use.

  • Simple set up, ideal for student use.
  • Contains everything needed to set up a distillation demonstration.
  • All glassware is made from high quality borosilicate glass.
  • Each set includes: 300 mm Liebig Condenser, 50 mm dia. Kjeldahl Ball, 500 mL Boiling Flask, 500 mL Erlenmeyer Flask, metal tripod stand, wire mesh gauze, cast iron support stand with rod, two prong clamp with bosshead.

Warranty

1 year limited manufacturer warranty.

Specifications

For Use With (Application) Distillation
Includes Liebig Condenser (30 mm), Kjeldahl Ball (50 mm), Boiling Flask (500 mL), Erlenmeyer Flask (500 mL) Support Stand, wire Gauze, Support Stand with Rod, Clamp with Bosshead
Material Borosilicate Glass, Metal, Rubber
Quantity 1 Ea.
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Sterility Non-sterile
Product Type Distillation Apparatus