missing translation for 'onlineSavingsMsg'
Learn More

Strontium Titanium Oxide Substrate, 5 x 5 x 1mm, Polished Two Sides, 110 Orientation, Thermo Scientific™

Catalog No. AA42671KT
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42671KT 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42671KT Supplier Thermo Scientific Chemicals Supplier No. 42671KT
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 12060-59-2
Molecular Formula O3SrTi
Molecular Weight (g/mol) 183.48
MDL Number MFCD00049554
InChI Key VEALVRVVWBQVSL-UHFFFAOYSA-N
Synonym strontium titanate, strontium titanium oxide, unii-olh4i98373, o3ti.sr, strontium titanium oxide srtio3, strontium titanium trioxide, titanate tio32-, strontium 1:1, strontium titanate 100g, strontium dioxido oxo titanium, strontium titanate, powder
PubChem CID 82899
IUPAC Name strontium;dioxido(oxo)titanium
SMILES [Sr++].[O-][Ti]([O-])=O

Specifications

Color White
Physical Form Substrate
Quantity 1 Ea.
Odor Odorless
Chemical Name or Material Strontium Titanium Oxide Substrate, Polished Two Sides
EINECSNumber 235-044-1
TSCA Yes
Recommended Storage Ambient temperatures