missing translation for 'onlineSavingsMsg'
Learn More

Strontium sulfate, Reagent Grade

Catalog No. AA3837322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3837322 100 g
AA3837314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3837322 Supplier Thermo Scientific Chemicals Supplier No. 03837322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7759-02-6 | O4SSr | 183.68 g/mol

Strontium sulfate is used as a precursor to other strontium compounds and as ceramic precursor. Its application in nitrate for use in pyrotechnics. Used as a fining agent in crystal glass. It is also used in powder coating, liquid paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7759-02-6

Specifications

Melting Point 1600°C
Physical Form Crystalline
Quantity 100 g
Molecular Formula O4SSr
MDL Number MFCD00036274
Merck Index 14,8848
Synonym strontium sulfate, strontium sulphate, sulfuric acid, strontium salt 1:1, strontium 2+ ion sulfate, unii-7q3kx2l47f, strontiumsulfat, celestine, strontium 2+ sulfate, strontium sulfate 25g
Solubility Information Soluble in dilute hydrochloric acid and nitric acid . Slightly soluble in water < 0.01g/L . Insoluble in ethanol and alkalis.
InChI Key UBXAKNTVXQMEAG-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)[O-].[Sr+2]
IUPAC Name strontium;sulfate
Molecular Weight (g/mol) 183.68
PubChem CID 3084026
Formula Weight 183.68
Grade Reagent
Odor Odorless
Density 3.960 g/mL
Chemical Name or Material Strontium sulfate, For reagent grade
Show More Show Less
EINECSNumber 231-850-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only