missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLSR23X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Sr | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Strontium Standard | |
| 7440-24-6 , 7697-37-2 , 7732-18-5 | |
| Sr | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| CIOAGBVUUVVLOB-UHFFFAOYSA-N | |
| 5% HNO3 | |
| 87.62 | |
| 5359327 | |
| Assurance |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP | |
| Certified Reference Material | |
| [Sr] | |
| strontium | |
| 500 mL | |
| CHEBI:33324 |
Chemical Identifiers
| 7440-24-6 | |
| 87.62 | |
| 5359327 | |
| strontium |
| Sr | |
| CIOAGBVUUVVLOB-UHFFFAOYSA-N | |
| CHEBI:33324 | |
| [Sr] |
Safety and Handling
Toxic in contact with skin. Causes serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only